Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, XBridge Peptide BEH C18, 300Å, 2.5 μm, 3 mm x 50 mm

Catalog No. 502391047 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XBridge Peptide BEH C18 XP Column, 300Å, 2.5 μm, 3 mm x 50 mm, is optimized and QC tested for peptide separations. It features a 300Å pore size trifunctionally bonded BEH particle, providing wide pH range, superior low pH stability, and ultra-low column bleed. Ideal for analyzing large peptides in proteomic, peptide mapping, and synthetic peptide applications. Each batch undergoes specific QC testing to ensure quality.

  • C18 chemistry, reversed phase
  • Temperature limits: 90 °C
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle size: 2.5 μm, pore size: 300 Å
  • Optimized and QC tested for peptide separations and analysis
  • Column format, BEH particle technology
  • System compatibility: HPLC, UHPLC
  • USP classification: L1
  • Dimensions: 3 mm inner diameter, 50 mm length

Catalog No. 50-239-1047 Supplier Waters Corp Supplier No. 186009071
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XBridge Peptide BEH C18 XP Column, 300Å, 2.5 μm, 3 mm x 50 mm, is optimized and QC tested for peptide separations. It features a 300Å pore size trifunctionally bonded BEH particle, providing wide pH range, superior low pH stability, and ultra-low column bleed. Ideal for analyzing large peptides in proteomic, peptide mapping, and synthetic peptide applications. Each batch undergoes specific QC testing to ensure quality.

  • C18 chemistry, reversed phase
  • Temperature limits: 90 °C
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle size: 2.5 μm, pore size: 300 Å
  • Optimized and QC tested for peptide separations and analysis
  • Column format, BEH particle technology
  • System compatibility: HPLC, UHPLC
  • USP classification: L1
  • Dimensions: 3 mm inner diameter, 50 mm length

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.