Learn More
Waters Corp WATERS CORPORATION XBridge BEH Phenyl Column, 130Å, 3.5 μm, 4.6 mm x 75 mm
Catalog No. 50781685 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH Phenyl column provides complementary selectivity to C18 stationary phases, offering ultra-low column bleed, long lifetimes, and excellent peak shape for HPLC applications. Crafted from 130Å porous silica with phenyl groups and 3.5 μm particles, it ensures superior resolution and efficient separation of polar and non-polar compounds. This column integrates seamlessly with various HPLC systems, providing stable, reproducible performance and a 12-month shelf life for diverse laboratory analyses.
  • Chemically stable phenyl sorbent for robust performance.
  • Ensures exceptional separation efficiency for diverse polar and non-polar compounds.
  • Designed with 130Å porous silica and 3.5 μm particles for superior peak resolution.
  • Integrates seamlessly with various HPLC systems and detectors.
  • Provides ease of use and maintenance for laboratory practicality.
Catalog No. 50-781-685 Supplier Waters Corp Supplier No. 186003333
The XBridge BEH Phenyl column provides complementary selectivity to C18 stationary phases, offering ultra-low column bleed, long lifetimes, and excellent peak shape for HPLC applications. Crafted from 130Å porous silica with phenyl groups and 3.5 μm particles, it ensures superior resolution and efficient separation of polar and non-polar compounds. This column integrates seamlessly with various HPLC systems, providing stable, reproducible performance and a 12-month shelf life for diverse laboratory analyses.
  • Chemically stable phenyl sorbent for robust performance.
  • Ensures exceptional separation efficiency for diverse polar and non-polar compounds.
  • Designed with 130Å porous silica and 3.5 μm particles for superior peak resolution.
  • Integrates seamlessly with various HPLC systems and detectors.
  • Provides ease of use and maintenance for laboratory practicality.