Learn More
Waters Corp XBridge Phenyl Column, 130Å, 3.5 μm, 2.1 mm x 50 mm
Catalog No. 50781674 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This chemically stable Phenyl sorbent offers ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Offers complementary selectivity for polyaromatic compounds to C18 stationary phases.
  • Ensures ultra-low column bleed, extended lifetimes, excellent peak shape, and high reproducibility.
  • Provides wide mobile phase pH and temperature flexibility (1 to 12 pH) for robust method development and UPLC transfer.
  • Features trifunctional bonding for exceptional low and high pH stability.
  • Maximum pressure 6000 psi (415 Bar); spherical particles with 3.5 μm size, 130 Å pore; 2.1 mm ID x 50 mm length.
Catalog No. 50-781-674 Supplier Waters Corp Supplier No. 186003322
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This chemically stable Phenyl sorbent offers ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Offers complementary selectivity for polyaromatic compounds to C18 stationary phases.
  • Ensures ultra-low column bleed, extended lifetimes, excellent peak shape, and high reproducibility.
  • Provides wide mobile phase pH and temperature flexibility (1 to 12 pH) for robust method development and UPLC transfer.
  • Features trifunctional bonding for exceptional low and high pH stability.
  • Maximum pressure 6000 psi (415 Bar); spherical particles with 3.5 μm size, 130 Å pore; 2.1 mm ID x 50 mm length.