Learn More
Waters Corp Waters XBridge BEH Phenyl Column, 3.5 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 186003335
Encompass_Preferred
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. These columns are the most chemically stable commercially available Phenyl sorbent on the market, providing ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Ultra-low column bleed
  • Long column lifetimes
  • Excellent peak shape
  • High and low pH stability due to innovative bonding and end-capping
  • Alternate selectivity for compounds with an aromatic moiety
  • Successful separation of aromatic acids with improved chemical stability
  • Flexibility with a wide range of mobile phase pH and temperature
  • Seamless method migration to UPLC technology
Catalog No. 50-781-687