Learn More
Waters Corp Waters XBridge BEH Phenyl Column, 130Å, 3.5 μm, 4.6 x 100 mm
Catalog No. 50781686 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge BEH Phenyl Column provides robust and reproducible chromatographic analysis, especially for compounds with aromatic rings. Its unique selectivity complements traditional C18 phases, and advanced bonding technology ensures high chemical stability, extended column life, and excellent peak performance. It supports a broad pH and temperature range, facilitating method development and UPLC system transfer.
  • Provides complementary selectivity to C18 stationary phases for polyaromatic compounds.
  • Ensures ultra-low column bleed, long lifetimes, and excellent peak shape.
  • Offers flexibility across a range of mobile phase pH and temperature.
  • Enables seamless method transfer to UPLC technology.
  • Features novel bonding and end-capping of BEH particle technology for stability.
  • Provides alternative selectivity for aromatic-containing compounds compared to straight-chain-alkyl columns.
Catalog No. 50-781-686 Supplier Waters Corp Supplier No. 186003334
This XBridge BEH Phenyl Column provides robust and reproducible chromatographic analysis, especially for compounds with aromatic rings. Its unique selectivity complements traditional C18 phases, and advanced bonding technology ensures high chemical stability, extended column life, and excellent peak performance. It supports a broad pH and temperature range, facilitating method development and UPLC system transfer.
  • Provides complementary selectivity to C18 stationary phases for polyaromatic compounds.
  • Ensures ultra-low column bleed, long lifetimes, and excellent peak shape.
  • Offers flexibility across a range of mobile phase pH and temperature.
  • Enables seamless method transfer to UPLC technology.
  • Features novel bonding and end-capping of BEH particle technology for stability.
  • Provides alternative selectivity for aromatic-containing compounds compared to straight-chain-alkyl columns.