Learn More
Waters Corp Waters XBridge BEH Phenyl Column, 5 μm, 2.1 mm ID, 100 mm Length
Catalog No. 50781691 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. Featuring a chemically stable Phenyl sorbent, this column provides ultra-low column bleed, long lifetimes, and excellent peak shape. It functions across a wide pH and temperature range, facilitating robust method development and easy transition to UPLC technology.
  • Provides complementary selectivity to C18 stationary phases for polyaromatic compounds.
  • Most chemically stable commercially available Phenyl sorbent on the market.
  • Offers ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Functions across a wide range of pH (1–12) and temperature.
  • Enables easy switching from conventional techniques to UPLC technology.
Catalog No. 50-781-691 Supplier Waters Corp Supplier No. 186003339
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. Featuring a chemically stable Phenyl sorbent, this column provides ultra-low column bleed, long lifetimes, and excellent peak shape. It functions across a wide pH and temperature range, facilitating robust method development and easy transition to UPLC technology.
  • Provides complementary selectivity to C18 stationary phases for polyaromatic compounds.
  • Most chemically stable commercially available Phenyl sorbent on the market.
  • Offers ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Functions across a wide range of pH (1–12) and temperature.
  • Enables easy switching from conventional techniques to UPLC technology.