Learn More
Waters Corp Waters Column, Phenyl, 5 μm, 2.1 mm x 50 mm
Catalog No. 50781690 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. These columns feature the most chemically stable commercially available Phenyl sorbent, resulting in ultra-low column bleed, extended column lifetimes, and excellent peak shape.
  • Complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Most chemically stable commercially available Phenyl sorbent.
  • Provides ultra-low column bleed.
  • Long column lifetimes.
  • Excellent peak shape.
  • Can withstand operating pressures up to 6000 psi (415 bar).
  • Functions within a pH range of 1 to 12.
Catalog No. 50-781-690 Supplier Waters Corp Supplier No. 186003338
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. These columns feature the most chemically stable commercially available Phenyl sorbent, resulting in ultra-low column bleed, extended column lifetimes, and excellent peak shape.
  • Complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Most chemically stable commercially available Phenyl sorbent.
  • Provides ultra-low column bleed.
  • Long column lifetimes.
  • Excellent peak shape.
  • Can withstand operating pressures up to 6000 psi (415 bar).
  • Functions within a pH range of 1 to 12.