Learn More
Waters Corp Waters XBridge BEH Phenyl Column, 130Å, 5 μm, 4.6 mm × 250 mm
Supplier:  Waters Corp 186003353
Encompass_Preferred
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. Featuring a chemically stable phenyl sorbent, these columns provide ultra-low bleed, long lifetimes, and excellent peak shape.
  • Offers alternate selectivity for compounds with an aromatic moiety.
  • Provides stability at low pH for enhanced chemical stability and column longevity.
  • BEH technology and particle synthesis enable scalability between HPLC and UPLC platforms.
  • Resistant to high pH dissolution, maintaining structural integrity and efficiency.
Catalog No. 50-781-705