Learn More
Waters Corp Waters XBridge BEH Phenyl Column, 130Å, 5 μm, 4.6 mm ID, 30 mm
Catalog No. 50781700 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns provide excellent performance for separating polyaromatic compounds. Designed with advanced Phenyl sorbent technology, these columns ensure reliable and consistent results for analytical applications.
  • Provides complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Most chemically stable commercially available Phenyl sorbent on the market.
  • Offers ultra-low column bleed.
  • Provides long column lifetimes.
  • Ensures excellent peak shape.
Catalog No. 50-781-700 Supplier Waters Corp Supplier No. 186003348
XBridge BEH Phenyl columns provide excellent performance for separating polyaromatic compounds. Designed with advanced Phenyl sorbent technology, these columns ensure reliable and consistent results for analytical applications.
  • Provides complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Most chemically stable commercially available Phenyl sorbent on the market.
  • Offers ultra-low column bleed.
  • Provides long column lifetimes.
  • Ensures excellent peak shape.