Learn More
Waters Corp XBridge BEH Phenyl Column, 130Å, 5 µm, 4.6 mm X 100 mm, 1/pk
Catalog No. 50781703 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.
Catalog No. 50-781-703 Supplier Waters Corp Supplier No. 186003351
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.