Learn More
Waters Corp Waters Corporation XBridge BEH Phenyl Column, 5 μm, 4.6 mm × 50 mm
Catalog No. 50781701 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds.
  • Features the most chemically stable commercially available Phenyl sorbent on the market.
  • Offers ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Operates effectively across a wide pH range of 1 to 12.
  • Sustains up to 6000 psi (415 Bar) pressure, assuming temperature and pH limits are observed.
  • Unique bonding and end-capping based on BEH particle technology enable remarkable low and high pH stability.
  • Most reproducible phenyl column available.
  • Suitable for smoothly transitioning methods to UPLC technology.
  • Phenyl hexyl-trifunctional ligand's bonding contributes to its stability.
Catalog No. 50-781-701 Supplier Waters Corp Supplier No. 186003349
  • XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds.
  • Features the most chemically stable commercially available Phenyl sorbent on the market.
  • Offers ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Operates effectively across a wide pH range of 1 to 12.
  • Sustains up to 6000 psi (415 Bar) pressure, assuming temperature and pH limits are observed.
  • Unique bonding and end-capping based on BEH particle technology enable remarkable low and high pH stability.
  • Most reproducible phenyl column available.
  • Suitable for smoothly transitioning methods to UPLC technology.
  • Phenyl hexyl-trifunctional ligand's bonding contributes to its stability.