Learn More
Waters Corp XBridge BEH Phenyl Analytical Column, 130 Å Pore, 3.5 μm Particle, 2.1 x 20 mm
Catalog No. 50781672 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH Phenyl Analytical Column is designed for analytical applications, featuring a 130 Å pore size and 3.5 μm particle size. It utilizes BEH Technology and Phenyl chemistry, providing robust performance for various chromatographic separations. This column measures 2.1 x 20 mm and is supplied as a single pack.
  • Utilizes BEH Technology (Ethylene Bridged Hybrid) synthesis.
  • Features Phenyl chemistry for selective separations.
  • Has a 130 Å pore diameter for optimal analyte diffusion.
  • Offers a 3.5 μm particle size for high resolution.
  • Measures 2.1 x 20 mm, suitable for efficient analysis.
  • Supplied as a single pack (1/pk).
  • Designed as an analytical column, not a guard column.
Catalog No. 50-781-672 Supplier Waters Corp Supplier No. 186003320
The XBridge BEH Phenyl Analytical Column is designed for analytical applications, featuring a 130 Å pore size and 3.5 μm particle size. It utilizes BEH Technology and Phenyl chemistry, providing robust performance for various chromatographic separations. This column measures 2.1 x 20 mm and is supplied as a single pack.
  • Utilizes BEH Technology (Ethylene Bridged Hybrid) synthesis.
  • Features Phenyl chemistry for selective separations.
  • Has a 130 Å pore diameter for optimal analyte diffusion.
  • Offers a 3.5 μm particle size for high resolution.
  • Measures 2.1 x 20 mm, suitable for efficient analysis.
  • Supplied as a single pack (1/pk).
  • Designed as an analytical column, not a guard column.