Learn More
Waters Corp XBridge BEH Phenyl Method Validation Kit, 130Å, 3.5 µm, 3 mm X 100 mm, 3/pk
Catalog No. 50465981 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.
Catalog No. 50-465-981 Supplier Waters Corp Supplier No. 186003800
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.