Learn More
Waters Corp Waters XBridge Premier Protein SEC Column, 250 Å, 2.5 μm, 4.6 x 150 mm
Catalog No. NC2239646 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier Protein SEC Column, 250Å, 2.5 μm, 4.6 x 150 mm, enables reliable protein size variant separations from 10,000 to approximately 650,000 Daltons. It uses MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies to reduce undesirable secondary interactions, simplifying method development with “generic” methods.
  • Designed for protein size variant separations (10,000 to 650,000 Daltons).
  • Features MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies.
  • Minimizes secondary ionic or hydrophobic interactions.
  • Supports “generic” method development.
  • Rigorously QC tested for consistent performance.
  • Applied in gel filtration, size exclusion, and analysis of aggregates, monomers, fragments, antibody conjugates.
This item is not returnable. View return policy
Catalog No. NC2239646 Supplier Waters Corp Supplier No. 186009959
The XBridge Premier Protein SEC Column, 250Å, 2.5 μm, 4.6 x 150 mm, enables reliable protein size variant separations from 10,000 to approximately 650,000 Daltons. It uses MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies to reduce undesirable secondary interactions, simplifying method development with “generic” methods.
  • Designed for protein size variant separations (10,000 to 650,000 Daltons).
  • Features MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies.
  • Minimizes secondary ionic or hydrophobic interactions.
  • Supports “generic” method development.
  • Rigorously QC tested for consistent performance.
  • Applied in gel filtration, size exclusion, and analysis of aggregates, monomers, fragments, antibody conjugates.