Learn More
Waters Corp Peptide Column, BEH C18, 130 Å, 2.5 μm, 4.6 x 150 mm
Catalog No. NC2895051 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is optimized and QC tested for peptide separations using cytochrome c digest standards. Its small-pore (130Å) trifunctionally bonded BEH particle ensures a wide usable pH range, superior low pH stability, and ultra-low column bleed. This improves reproducibility, peak shape, and recovery for metal-sensitive analytes in proteomics, protein characterization, and peptide synthesis.
  • Features small-pore (130Å) trifunctionally bonded BEH particle.
  • Offers wide usable pH range: 1 pH to 12 pH.
  • Provides superior low pH stability.
  • Ensures ultra-low column bleed for assaying samples.
  • Improves reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Ideal for proteomics, protein characterization, peptide synthesis, and peptide analysis.
Catalog No. NC2895051 Supplier Waters Corp Supplier No. 186009900
This column is optimized and QC tested for peptide separations using cytochrome c digest standards. Its small-pore (130Å) trifunctionally bonded BEH particle ensures a wide usable pH range, superior low pH stability, and ultra-low column bleed. This improves reproducibility, peak shape, and recovery for metal-sensitive analytes in proteomics, protein characterization, and peptide synthesis.
  • Features small-pore (130Å) trifunctionally bonded BEH particle.
  • Offers wide usable pH range: 1 pH to 12 pH.
  • Provides superior low pH stability.
  • Ensures ultra-low column bleed for assaying samples.
  • Improves reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Ideal for proteomics, protein characterization, peptide synthesis, and peptide analysis.