Learn More
Waters Corp Waters Corporation XBridge Premier BEH C18 Column, 130Å, 2.5 μm, 2.1 x 150 mm
Catalog No. 502224250 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is ideal for method development, operating across a wide mobile phase pH range (1 to 12). It features a trifunctionally bonded C18 ligand, making it a universal column for most LC separations. It delivers superior stability, ultra-low column bleed, and exceptional performance for UHPLC and HPLC assays. MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Wide usable pH range (1 to 12) for versatile method development.
  • BEH particle technology ensures enhanced stability.
  • MaxPeak High Performance Surface minimizes analyte-surface interactions.
  • Exceptional performance and throughput for UHPLC/HPLC assays.
  • Trifunctionally bonded C18 ligand for universal LC applicability.
  • Improved reproducibility and efficiency via BEH particle.
  • Seamless UPLC transition, VanGuard FIT compatible.
Catalog No. 50-222-4250 Supplier Waters Corp Supplier No. 186009829
This column is ideal for method development, operating across a wide mobile phase pH range (1 to 12). It features a trifunctionally bonded C18 ligand, making it a universal column for most LC separations. It delivers superior stability, ultra-low column bleed, and exceptional performance for UHPLC and HPLC assays. MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Wide usable pH range (1 to 12) for versatile method development.
  • BEH particle technology ensures enhanced stability.
  • MaxPeak High Performance Surface minimizes analyte-surface interactions.
  • Exceptional performance and throughput for UHPLC/HPLC assays.
  • Trifunctionally bonded C18 ligand for universal LC applicability.
  • Improved reproducibility and efficiency via BEH particle.
  • Seamless UPLC transition, VanGuard FIT compatible.