Learn More
Waters Corp Column, BEH C18, 3.5 μm, 2.1 x 150 mm
Catalog No. 502886192 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Premier BEH C18 sorbent is designed for method development and versatile liquid chromatography across a wide mobile phase pH range (1 to 12). Its trifunctionally bonded C18 ligand ensures superior low pH stability and minimal column bleed. MaxPeak High Performance Surface technology minimizes unwanted analyte interactions, enhancing peak shape.
  • Versatile liquid chromatography separations (pH 1 to 12).
  • Superior low pH stability and ultra-low column bleed.
  • Exceptional HPLC performance, enabling seamless UPLC transition.
  • MaxPeak High Performance Surface reduces unwanted analyte interactions.
Catalog No. 50-288-6192 Supplier Waters Corp Supplier No. 186010652
The Premier BEH C18 sorbent is designed for method development and versatile liquid chromatography across a wide mobile phase pH range (1 to 12). Its trifunctionally bonded C18 ligand ensures superior low pH stability and minimal column bleed. MaxPeak High Performance Surface technology minimizes unwanted analyte interactions, enhancing peak shape.
  • Versatile liquid chromatography separations (pH 1 to 12).
  • Superior low pH stability and ultra-low column bleed.
  • Exceptional HPLC performance, enabling seamless UPLC transition.
  • MaxPeak High Performance Surface reduces unwanted analyte interactions.