Learn More
Waters Corp Column, BEH C18, 3.5 μm, 4.6 x 250 mm
Catalog No. 502886206 Shop All Waters Corporation Products
Encompass_Preferred
This XBridge Premier BEH C18 sorbent column is designed for method development, usable across a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand offers a universal column for most LC separations, providing the widest usable pH range, superior low pH stability, and ultra-low column bleed. XBridge Premier 3.5 μm Columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, facilitating UPLC transition. MaxPeak High Performance Surface hardware significantly reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss.
  • Offers the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed.
  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Reduces unwanted analyte/surface interactions with MaxPeak High Performance Surface technology.
Catalog No. 50-288-6206 Supplier Waters Corp Supplier No. 186010662
This XBridge Premier BEH C18 sorbent column is designed for method development, usable across a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand offers a universal column for most LC separations, providing the widest usable pH range, superior low pH stability, and ultra-low column bleed. XBridge Premier 3.5 μm Columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, facilitating UPLC transition. MaxPeak High Performance Surface hardware significantly reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss.
  • Offers the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed.
  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Reduces unwanted analyte/surface interactions with MaxPeak High Performance Surface technology.