Learn More
Waters Corp XBridge Premier BEH C18 Column, 3.5 μm, 4.6 × 50 mm
Catalog No. 502886188 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge Premier BEH C18 sorbent column, 3.5 μm, 4.6 × 50 mm, is for method development across a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand provides versatility for LC separations with excellent low pH stability and minimal column bleed. It offers superior HPLC assay performance and a smooth transition to UPLC, incorporating MaxPeak High Performance Surface technology to reduce analyte/surface interactions and improve peak shape.
  • MaxPeak High Performance Surface hardware technology reduces analyte/surface interactions.
  • Trifunctionally bonded C18 ligand for universal LC separations.
  • Widest usable pH range (1-12).
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.
Catalog No. 50-288-6188 Supplier Waters Corp Supplier No. 186010658
This XBridge Premier BEH C18 sorbent column, 3.5 μm, 4.6 × 50 mm, is for method development across a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand provides versatility for LC separations with excellent low pH stability and minimal column bleed. It offers superior HPLC assay performance and a smooth transition to UPLC, incorporating MaxPeak High Performance Surface technology to reduce analyte/surface interactions and improve peak shape.
  • MaxPeak High Performance Surface hardware technology reduces analyte/surface interactions.
  • Trifunctionally bonded C18 ligand for universal LC separations.
  • Widest usable pH range (1-12).
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.