Learn More
Waters Corp XBridge Premier BEH C18 Column, 3.5 μm, 4.6 mm ID, 100 mm Length
Catalog No. 502886220 Shop All Waters Corporation Products
Encompass_Preferred
The XBridge Premier BEH C18 sorbent in the 3.5 μm, 4.6 x 100 mm column is a universal solution for LC separations, usable across a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand ensures superior low pH stability and ultra-low column bleed. MaxPeak High Performance Surface hardware technology minimizes unwanted analyte/surface interactions, leading to improved peak shape and signal intensity.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity, peak shapes, and recovery.
  • Good universal column choice for a wide variety of compounds.
  • Stable across a wide pH range and at high temperatures.
Catalog No. 50-288-6220 Supplier Waters Corp Supplier No. 186010660
The XBridge Premier BEH C18 sorbent in the 3.5 μm, 4.6 x 100 mm column is a universal solution for LC separations, usable across a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand ensures superior low pH stability and ultra-low column bleed. MaxPeak High Performance Surface hardware technology minimizes unwanted analyte/surface interactions, leading to improved peak shape and signal intensity.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity, peak shapes, and recovery.
  • Good universal column choice for a wide variety of compounds.
  • Stable across a wide pH range and at high temperatures.