Learn More
Waters Corp Column, C18, 3.5 μm, 4.6 mm, 150 mm
Catalog No. 502886238 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This BEH C18 column (3.5 μm, 4.6 x 150 mm) is engineered for method development, supporting a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand ensures broad applicability for LC separations, offering a wide usable pH range, excellent low pH stability, and ultra-low column bleed. Integrated MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal intensity reductions.
  • Supports method development across mobile phase pH range of 1 to 12.
  • Trifunctionally bonded C18 ligand for universal LC separations.
  • Features wide pH range, excellent low pH stability, and ultra-low column bleed.
  • Delivers exceptional performance, robustness, and throughput for HPLC.
  • Enables smooth transition to UPLC adoption.
  • MaxPeak High Performance Surface hardware reduces analyte/surface interactions.
Catalog No. 50-288-6238 Supplier Waters Corp Supplier No. 186010661
This BEH C18 column (3.5 μm, 4.6 x 150 mm) is engineered for method development, supporting a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand ensures broad applicability for LC separations, offering a wide usable pH range, excellent low pH stability, and ultra-low column bleed. Integrated MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal intensity reductions.
  • Supports method development across mobile phase pH range of 1 to 12.
  • Trifunctionally bonded C18 ligand for universal LC separations.
  • Features wide pH range, excellent low pH stability, and ultra-low column bleed.
  • Delivers exceptional performance, robustness, and throughput for HPLC.
  • Enables smooth transition to UPLC adoption.
  • MaxPeak High Performance Surface hardware reduces analyte/surface interactions.