Learn More
Waters Corp Waters Columns, C18, 130Å, 2.5 μm, 2.1 mm x 100 mm
Catalog No. 502224249 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This C18 sorbent, ideal for method development, can be used over a mobile phase pH range of 1-12. It serves as a universal column for most LC separations, offering superior low pH stability and ultra-low column bleed.
  • Provides a universal column for most LC separations with a trifunctionally bonded C18 ligand.
  • Ensures exceptional separation performance, robustness, and throughput for UHPLC and HPLC assays with 2.5 μm particles.
  • Enables a seamless transition path toward UPLC adoption.
  • Features MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions.
  • Utilizes an Ethylene-Bridged Hybrid (BEH) particle for the highest chromatographic particle stability.
  • Withstands pressures up to 18,000 PSI.
Catalog No. 50-222-4249 Supplier Waters Corp Supplier No. 186009828
This C18 sorbent, ideal for method development, can be used over a mobile phase pH range of 1-12. It serves as a universal column for most LC separations, offering superior low pH stability and ultra-low column bleed.
  • Provides a universal column for most LC separations with a trifunctionally bonded C18 ligand.
  • Ensures exceptional separation performance, robustness, and throughput for UHPLC and HPLC assays with 2.5 μm particles.
  • Enables a seamless transition path toward UPLC adoption.
  • Features MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions.
  • Utilizes an Ethylene-Bridged Hybrid (BEH) particle for the highest chromatographic particle stability.
  • Withstands pressures up to 18,000 PSI.