Learn More
Waters Corp XBridge Premier BEH C18 VanGuard FIT Column, 3.5 μm, 2.1 x 50 mm
Catalog No. 502886180 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This robust column for method development uses BEH C18 sorbent, operating across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand suits most LC separations with broad pH compatibility, excellent low pH stability, and minimal column bleed. It delivers high performance for HPLC assays and facilitates UPLC transition. MaxPeak High Performance Surface hardware minimizes analyte interactions, improving peak shape and maintaining signal intensity.
  • Operates across a mobile phase pH range of 1 to 12.
  • Trifunctionally bonded C18 ligand ensures broad pH compatibility and superior low pH stability.
  • MaxPeak High Performance Surface technology minimizes analyte interactions, improving peak shape and signal intensity.
  • Enables seamless transition from HPLC to UPLC.
  • Increases analyte recovery, sensitivity, and reproducibility; reduces column conditioning.
Catalog No. 50-288-6180 Supplier Waters Corp Supplier No. 186010646
This robust column for method development uses BEH C18 sorbent, operating across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand suits most LC separations with broad pH compatibility, excellent low pH stability, and minimal column bleed. It delivers high performance for HPLC assays and facilitates UPLC transition. MaxPeak High Performance Surface hardware minimizes analyte interactions, improving peak shape and maintaining signal intensity.
  • Operates across a mobile phase pH range of 1 to 12.
  • Trifunctionally bonded C18 ligand ensures broad pH compatibility and superior low pH stability.
  • MaxPeak High Performance Surface technology minimizes analyte interactions, improving peak shape and signal intensity.
  • Enables seamless transition from HPLC to UPLC.
  • Increases analyte recovery, sensitivity, and reproducibility; reduces column conditioning.