Learn More
Waters Corp XBridge Premier BEH C18 VanGuard FIT Column, 3.5 μm, 4.6 mm ID, 250 mm Length
Catalog No. 502886187 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier BEH C18 sorbent, with a 3.5 μm particle size and dimensions of 4.6 x 250 mm, functions as a workhorse for method development and can be utilized across a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand offers a universal column suitable for most LC separations, providing the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Delivers exceptional separation performance, robustness, and throughput for HPLC assays, while also enabling a smooth transition to UPLC adoption.
  • Incorporates MaxPeak High Performance Surface hardware technology, which significantly lessens unwanted analyte/surface interactions that can cause poor peak shape and reductions in signal intensity.
Catalog No. 50-288-6187 Supplier Waters Corp Supplier No. 186010656
The XBridge Premier BEH C18 sorbent, with a 3.5 μm particle size and dimensions of 4.6 x 250 mm, functions as a workhorse for method development and can be utilized across a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand offers a universal column suitable for most LC separations, providing the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Delivers exceptional separation performance, robustness, and throughput for HPLC assays, while also enabling a smooth transition to UPLC adoption.
  • Incorporates MaxPeak High Performance Surface hardware technology, which significantly lessens unwanted analyte/surface interactions that can cause poor peak shape and reductions in signal intensity.