Learn More
Waters Corp Column, 3.5 μm Particle, 4.6 x 150 mm, BEH C18
Catalog No. 502886242 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge Premier BEH C18 column, with 3.5 μm particle size and 4.6 x 150 mm dimensions, is optimized for method development. It provides a wide mobile phase pH range (1–12), superior low pH stability, and ultra-low column bleed. MaxPeak High Performance Surface hardware minimizes analyte/surface interactions, enhancing peak shape.
  • Universal column for LC separations, stable across a wide pH range (1–12) and high temperatures.
  • Superior low pH stability and ultra-low column bleed.
  • Exceptional separation performance, robustness, and throughput for HPLC assays, with seamless UPLC transition.
  • MaxPeak High Performance Surface hardware reduces analyte/surface interactions, minimizing loss and improving peak shapes, and sensitivity.
  • Reduces column conditioning and passivation times.
  • Integrated guard column design maintains separation efficiency and lifetime.
Catalog No. 50-288-6242 Supplier Waters Corp Supplier No. 186010655
This XBridge Premier BEH C18 column, with 3.5 μm particle size and 4.6 x 150 mm dimensions, is optimized for method development. It provides a wide mobile phase pH range (1–12), superior low pH stability, and ultra-low column bleed. MaxPeak High Performance Surface hardware minimizes analyte/surface interactions, enhancing peak shape.
  • Universal column for LC separations, stable across a wide pH range (1–12) and high temperatures.
  • Superior low pH stability and ultra-low column bleed.
  • Exceptional separation performance, robustness, and throughput for HPLC assays, with seamless UPLC transition.
  • MaxPeak High Performance Surface hardware reduces analyte/surface interactions, minimizing loss and improving peak shapes, and sensitivity.
  • Reduces column conditioning and passivation times.
  • Integrated guard column design maintains separation efficiency and lifetime.