Learn More
Waters Corp VanGuard Guards, BEH C8, 1.7 μm, 2.1 x 5 mm
Catalog No. 502269370 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier Cartridges are designed to work with ACQUITY Premier VanGuard FIT columns. These integrated guard cartridges serve to protect the analytical column from matrix and sample contaminants, thereby extending the column's lifetime.
  • Designed for use with compatible columns.
  • Fully integrated guard cartridges.
  • Protects the analytical column from matrix and sample contaminants.
  • Extends column lifetime.
This item is not returnable. View return policy
Catalog No. 50-226-9370 Supplier Waters Corp Supplier No. 186010362
ACQUITY Premier Cartridges are designed to work with ACQUITY Premier VanGuard FIT columns. These integrated guard cartridges serve to protect the analytical column from matrix and sample contaminants, thereby extending the column's lifetime.
  • Designed for use with compatible columns.
  • Fully integrated guard cartridges.
  • Protects the analytical column from matrix and sample contaminants.
  • Extends column lifetime.