Learn More
Waters Corp Waters XBridge Premier BEH C8 VanGuard FIT Column, 2.5 μm, 4.6 mm, 50 mm
Catalog No. 502269381 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed for high-performance liquid chromatography, offering reduced retention and a wide pH range. They feature MaxPeak High Performance Surface technology and a VanGuard Fit design for enhanced performance and longevity.
  • Reduced retention compared to C18 columns, suitable for fast methods development.
  • Wide pH range of 1–12 provided by trifunctionally bonded BEH particles.
  • Superior low pH stability.
  • Ultra-low column bleed.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime.
This item is not returnable. View return policy
Catalog No. 50-226-9381 Supplier Waters Corp Supplier No. 186010373
These columns are designed for high-performance liquid chromatography, offering reduced retention and a wide pH range. They feature MaxPeak High Performance Surface technology and a VanGuard Fit design for enhanced performance and longevity.
  • Reduced retention compared to C18 columns, suitable for fast methods development.
  • Wide pH range of 1–12 provided by trifunctionally bonded BEH particles.
  • Superior low pH stability.
  • Ultra-low column bleed.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime.