Learn More
Waters Corp Waters XBridge Premier BEH C8 VanGuard FIT Column, 2.5 μm, 4.6 mm X 150 mm
Catalog No. 502269383 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier BEH C8 Columns provide less retention than C18 columns, facilitating rapid method development. They leverage trifunctionally bonded BEH particles for a broad pH range (1 to 12), excellent low pH stability, and minimal column bleed, alongside MaxPeak High Performance Surface hardware to enhance peak shape and signal intensity. The VanGuard Fit design extends column lifespan by protecting against contaminants.
  • Less retentive compared to C18 columns, which is desirable for fast method development.
  • Trifunctionally bonded BEH particles offer the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime.
This item is not returnable. View return policy
Catalog No. 50-226-9383 Supplier Waters Corp Supplier No. 186010375
The XBridge Premier BEH C8 Columns provide less retention than C18 columns, facilitating rapid method development. They leverage trifunctionally bonded BEH particles for a broad pH range (1 to 12), excellent low pH stability, and minimal column bleed, alongside MaxPeak High Performance Surface hardware to enhance peak shape and signal intensity. The VanGuard Fit design extends column lifespan by protecting against contaminants.
  • Less retentive compared to C18 columns, which is desirable for fast method development.
  • Trifunctionally bonded BEH particles offer the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime.