Learn More
Waters Corp Waters XBridge Premier BEH C8 Column, 2.5 μm, 4.6 × 150 mm
Catalog No. 502269380 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier BEH C8 Column is engineered for UHPLC/UPLC, offering less retention than C18 columns for rapid method development. It integrates trifunctionally bonded BEH particles for a wide pH range (1-12) and MaxPeak High Performance Surface technology to minimize analyte/surface interactions, enhancing peak shape and signal.
  • Less retentive than C18 columns, ideal for rapid method development.
  • Wide usable pH range (1 to 12) with superior low pH stability and ultra-low column bleed.
  • MaxPeak High Performance Surface technology reduces analyte/surface interactions, increasing recovery, sensitivity, and reproducibility.
  • Reduces conditioning times and improves peak shapes.
  • Good universal choice, stable across wide pH and high temperatures, simplifying mobile phases.
This item is not returnable. View return policy
Catalog No. 50-226-9380 Supplier Waters Corp Supplier No. 186010372
The XBridge Premier BEH C8 Column is engineered for UHPLC/UPLC, offering less retention than C18 columns for rapid method development. It integrates trifunctionally bonded BEH particles for a wide pH range (1-12) and MaxPeak High Performance Surface technology to minimize analyte/surface interactions, enhancing peak shape and signal.
  • Less retentive than C18 columns, ideal for rapid method development.
  • Wide usable pH range (1 to 12) with superior low pH stability and ultra-low column bleed.
  • MaxPeak High Performance Surface technology reduces analyte/surface interactions, increasing recovery, sensitivity, and reproducibility.
  • Reduces conditioning times and improves peak shapes.
  • Good universal choice, stable across wide pH and high temperatures, simplifying mobile phases.