Learn More
Waters Corp XBridge Premier BEH C8 VanGuard FIT Column, MaxPeak HPS, 2.5 μm, 4.6 x 100 mm
Catalog No. 502269382 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Premier BEH C8 VanGuard FIT Columns are engineered with BEH Technology™ and MaxPeak High Performance Surfaces (HPS) to optimize performance in high-speed applications. These columns are designed to enhance analyte recovery, sensitivity, and reproducibility by minimizing unwanted interactions between analytes and surfaces.
  • Increase analyte recovery, sensitivity, and reproducibility
  • Minimize analyte/surface interactions that can lead to sample losses
  • Mitigate loss of metal-sensitive analytes like lipids, organic acids, and peptides
  • Reduce column conditioning and passivation times
  • Improve sensitivity and peak shapes
  • Enable simpler mobile phases, without complex additives
  • Save time in method development
  • Provide greater confidence in data and decision-making
This item is not returnable. View return policy
Catalog No. 50-226-9382 Supplier Waters Corp Supplier No. 186010374
XBridge Premier BEH C8 VanGuard FIT Columns are engineered with BEH Technology™ and MaxPeak High Performance Surfaces (HPS) to optimize performance in high-speed applications. These columns are designed to enhance analyte recovery, sensitivity, and reproducibility by minimizing unwanted interactions between analytes and surfaces.
  • Increase analyte recovery, sensitivity, and reproducibility
  • Minimize analyte/surface interactions that can lead to sample losses
  • Mitigate loss of metal-sensitive analytes like lipids, organic acids, and peptides
  • Reduce column conditioning and passivation times
  • Improve sensitivity and peak shapes
  • Enable simpler mobile phases, without complex additives
  • Save time in method development
  • Provide greater confidence in data and decision-making