Learn More
Waters Corp Waters XBridge Premier BEH C8 Column, 2.5 μm, 2.1 x 50 mm
Catalog No. 502269374 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier BEH C8 VanGuard FIT Column features a 2.5 μm particle size and 2.1 x 50 mm dimensions. It is designed for high-performance separations, leveraging advanced technology to improve analytical results.
  • Reduces column conditioning and passivation times.
  • Improves sensitivity and peak shapes.
  • Enables simpler mobile phases without complex additives.
  • Utilizes high performance surfaces technology to minimize analyte/surface interactions, enhancing recovery, sensitivity, and reproducibility.
  • Offers broad applicability for small molecule, peptide, oligonucleotide, and glycan separations.
  • Provides a general purpose phase for method development due to pH and temperature stability.
  • Ideal for analyzing strongly hydrophobic compounds.
  • Extends column lifetime and maintains separation efficiency through integrated guard column technology.
This item is not returnable. View return policy
Catalog No. 50-226-9374 Supplier Waters Corp Supplier No. 186010366
The XBridge Premier BEH C8 VanGuard FIT Column features a 2.5 μm particle size and 2.1 x 50 mm dimensions. It is designed for high-performance separations, leveraging advanced technology to improve analytical results.
  • Reduces column conditioning and passivation times.
  • Improves sensitivity and peak shapes.
  • Enables simpler mobile phases without complex additives.
  • Utilizes high performance surfaces technology to minimize analyte/surface interactions, enhancing recovery, sensitivity, and reproducibility.
  • Offers broad applicability for small molecule, peptide, oligonucleotide, and glycan separations.
  • Provides a general purpose phase for method development due to pH and temperature stability.
  • Ideal for analyzing strongly hydrophobic compounds.
  • Extends column lifetime and maintains separation efficiency through integrated guard column technology.