Learn More
Waters Corp Waters XBridge Premier BEH HILIC Column, 2.5 μm, 4.6 × 100 mm
Catalog No. 502269400 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XBridge Premier BEH HILIC Columns retain and separate polar compounds using unbonded BEH (Bridged Ethyl Hybrid) particles. Optimized for efficient, reproducible separations across a wide pH range, they offer greater ruggedness and longer column lifetimes than silica-based HILIC phases. MaxPeak High Performance Surface hardware reduces unwanted analyte interactions, preventing poor peak shape and signal loss.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested for efficient, reproducible separations across a wide pH range.
  • More rugged than unbonded silica-based HILIC phases.
  • BEH particle's chemical stability results in long column lifetimes.
  • Utilizes MaxPeak High Performance Surface hardware technology.
  • Significantly reduces unwanted analyte/surface interactions.
  • Prevents poor peak shape and losses in signal intensity.
This item is not returnable. View return policy
Catalog No. 50-226-9400 Supplier Waters Corp Supplier No. 186010392
Add to Cart
Add to Cart
These XBridge Premier BEH HILIC Columns retain and separate polar compounds using unbonded BEH (Bridged Ethyl Hybrid) particles. Optimized for efficient, reproducible separations across a wide pH range, they offer greater ruggedness and longer column lifetimes than silica-based HILIC phases. MaxPeak High Performance Surface hardware reduces unwanted analyte interactions, preventing poor peak shape and signal loss.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested for efficient, reproducible separations across a wide pH range.
  • More rugged than unbonded silica-based HILIC phases.
  • BEH particle's chemical stability results in long column lifetimes.
  • Utilizes MaxPeak High Performance Surface hardware technology.
  • Significantly reduces unwanted analyte/surface interactions.
  • Prevents poor peak shape and losses in signal intensity.