Learn More
Waters Corp Waters VanGuard FIT Cartridge, BEH HILIC, 2.5 μm, 2.1 x 5 mm
Supplier:  Waters Corp 186010390
Encompass_Preferred
XBridge Premier Cartridges are designed to work with XBridge Premier VanGuard FIT columns. These fully integrated guard cartridges protect the analytical column from matrix and sample contaminants, thereby extending column lifetime.
  • Protects the analytical column from matrix and sample contaminants.
  • Extends column lifetime.
Catalog No. 50-226-9398