Learn More
Waters Corp Waters VanGuard FIT Cartridge, 1.7 μm, 2.1 x 5 mm, HILIC, BEH
Catalog No. 502269391 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier Cartridges work with the ACQUITY Premier VanGuard FIT columns. These fully integrated guard cartridges protect the analytical column from matrix and sample contaminants, extending column lifetime.
  • Designed to work with compatible VanGuard FIT columns.
  • Fully integrated guard cartridges.
  • Protects the analytical column from matrix and sample contaminants.
  • Extends column lifetime.
This item is not returnable. View return policy
Catalog No. 50-226-9391 Supplier Waters Corp Supplier No. 186010383
ACQUITY Premier Cartridges work with the ACQUITY Premier VanGuard FIT columns. These fully integrated guard cartridges protect the analytical column from matrix and sample contaminants, extending column lifetime.
  • Designed to work with compatible VanGuard FIT columns.
  • Fully integrated guard cartridges.
  • Protects the analytical column from matrix and sample contaminants.
  • Extends column lifetime.