Learn More
Waters Corp Waters Corporation XBridge Premier BEH HILIC VanGuard FIT Column, 2.5 μm, 2.1 mm, 150 mm
Catalog No. 502269397 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns feature unbonded BEH (Bridged Ethyl Hybrid) particles for retaining and separating highly polar compounds. Optimized for efficient, reproducible separations across a broad pH range, their chemical stability provides greater robustness than silica-based HILIC phases, extending column lifetimes. MaxPeak High Performance Surface hardware minimizes analyte/surface interactions, preventing poor peak shape and signal loss. The VanGuard Fit design also protects against contaminants, increasing lifespan.
  • Designed for retaining and separating very polar compounds.
  • Optimized for efficient, reproducible separations across a wide pH range.
  • BEH particle's chemical stability results in long column lifetimes.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions.
  • VanGuard Fit column design protects from contaminants and increases column lifetime.
This item is not returnable. View return policy
Catalog No. 50-226-9397 Supplier Waters Corp Supplier No. 186010389
These columns feature unbonded BEH (Bridged Ethyl Hybrid) particles for retaining and separating highly polar compounds. Optimized for efficient, reproducible separations across a broad pH range, their chemical stability provides greater robustness than silica-based HILIC phases, extending column lifetimes. MaxPeak High Performance Surface hardware minimizes analyte/surface interactions, preventing poor peak shape and signal loss. The VanGuard Fit design also protects against contaminants, increasing lifespan.
  • Designed for retaining and separating very polar compounds.
  • Optimized for efficient, reproducible separations across a wide pH range.
  • BEH particle's chemical stability results in long column lifetimes.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions.
  • VanGuard Fit column design protects from contaminants and increases column lifetime.