Learn More
Waters Corp XBridge Premier BEH Phenyl Column, 3.5 μm, 4.6 x 50 mm
Catalog No. 502886183 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XBridge Premier BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. The sorbent provides the most chemically stable phenyl sorbent on the market, ensuring ultra-low column bleed, long column lifetimes, and excellent peak shape. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity.
  • Complementary selectivity to C18 stationary phases for polyaromatic compounds.
  • Chemically stable phenyl sorbent.
  • Ultra-low column bleed.
  • Long column lifetimes.
  • Excellent peak shape.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • Prevents poor peak shape and signal intensity losses.
Catalog No. 50-288-6183 Supplier Waters Corp Supplier No. 186010675
These XBridge Premier BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. The sorbent provides the most chemically stable phenyl sorbent on the market, ensuring ultra-low column bleed, long column lifetimes, and excellent peak shape. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity.
  • Complementary selectivity to C18 stationary phases for polyaromatic compounds.
  • Chemically stable phenyl sorbent.
  • Ultra-low column bleed.
  • Long column lifetimes.
  • Excellent peak shape.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • Prevents poor peak shape and signal intensity losses.