Learn More
Waters Corp Column, XBridge Premier BEH Phenyl, 3.5 μm, 4.6 x 250 mm
Catalog No. 502886236 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Premier BEH Phenyl 3.5 μm, 4.6 x 250 mm Columns provide complementary selectivity to C18 phases, especially for polyaromatic compounds. Featuring a highly stable phenyl sorbent, these columns ensure ultra-low bleed, extended lifetimes, and excellent peak shape. MaxPeak Premier hardware reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss.
  • Complementary selectivity to C18 stationary phases
  • Especially effective for polyaromatic compounds
  • Most chemically stable phenyl sorbent on the market
  • Ultra-low column bleed
  • Long column lifetimes
  • Excellent peak shape
  • MaxPeak High Performance Surface hardware technology for reduced unwanted analyte/surface interactions
Catalog No. 50-288-6236 Supplier Waters Corp Supplier No. 186010678
XBridge Premier BEH Phenyl 3.5 μm, 4.6 x 250 mm Columns provide complementary selectivity to C18 phases, especially for polyaromatic compounds. Featuring a highly stable phenyl sorbent, these columns ensure ultra-low bleed, extended lifetimes, and excellent peak shape. MaxPeak Premier hardware reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss.
  • Complementary selectivity to C18 stationary phases
  • Especially effective for polyaromatic compounds
  • Most chemically stable phenyl sorbent on the market
  • Ultra-low column bleed
  • Long column lifetimes
  • Excellent peak shape
  • MaxPeak High Performance Surface hardware technology for reduced unwanted analyte/surface interactions