Learn More
Waters Corp XBridge Premier BEH Phenyl Column, 130 Å, 3.5 μm
Catalog No. 502886204 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge Premier BEH Phenyl VanGuard FIT Column is designed for high-performance liquid chromatography applications. It incorporates MaxPeak High Performance Surfaces technology to provide enhanced analytical performance and extended column life.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Improved recovery.
  • Simpler mobile phases, without complex additives.
  • Time savings in method development.
  • Reduced risk and greater confidence in data and decision making.
  • Integrated guard column technology protects investment.
  • Maintains separation efficiency.
  • Extends column lifetime.
Catalog No. 50-288-6204 Supplier Waters Corp Supplier No. 186010670
This XBridge Premier BEH Phenyl VanGuard FIT Column is designed for high-performance liquid chromatography applications. It incorporates MaxPeak High Performance Surfaces technology to provide enhanced analytical performance and extended column life.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Improved recovery.
  • Simpler mobile phases, without complex additives.
  • Time savings in method development.
  • Reduced risk and greater confidence in data and decision making.
  • Integrated guard column technology protects investment.
  • Maintains separation efficiency.
  • Extends column lifetime.