Learn More
Waters Corp XBridge Premier BEH Phenyl VanGuard FIT Column, 3.5 μm, 4.6 x 250 mm
Catalog No. 502886186 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier BEH Phenyl VanGuard FIT Column integrates Ethylene Bridged Hybrid (BEH) particles with MaxPeak High Performance Surfaces (HPS) Technology. This enhances chromatographic performance by optimizing analyte peak shape and improving recovery, sensitivity, and reproducibility across a wide range of temperatures and pH.
  • MaxPeak HPS technology prevents undesired analyte/surface interactions.
  • Improved analyte sensitivity and peak shapes.
  • Reduced column conditioning for higher productivity.
  • High pH stability (1 to 12) and chemical resistance.
  • BEH technology offers improved peak shape for bases.
  • Reproducible phenyl chemistry for alternative selectivity.
  • Ideal for polyaromatic compound separation.
  • VanGuard FIT for integrated, cost-effective column protection.
Catalog No. 50-288-6186 Supplier Waters Corp Supplier No. 186010673
The XBridge Premier BEH Phenyl VanGuard FIT Column integrates Ethylene Bridged Hybrid (BEH) particles with MaxPeak High Performance Surfaces (HPS) Technology. This enhances chromatographic performance by optimizing analyte peak shape and improving recovery, sensitivity, and reproducibility across a wide range of temperatures and pH.
  • MaxPeak HPS technology prevents undesired analyte/surface interactions.
  • Improved analyte sensitivity and peak shapes.
  • Reduced column conditioning for higher productivity.
  • High pH stability (1 to 12) and chemical resistance.
  • BEH technology offers improved peak shape for bases.
  • Reproducible phenyl chemistry for alternative selectivity.
  • Ideal for polyaromatic compound separation.
  • VanGuard FIT for integrated, cost-effective column protection.