Learn More
Waters Corp XBridge Premier BEH Phenyl VanGuard FIT Column, 3.5 μm, 2.1 x 100 mm
Catalog No. 502886190 Shop All Waters Corporation Products
Encompass_Preferred
This Premier BEH Phenyl VanGuard FIT Column provides complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. Its sorbent offers high chemical stability, ensuring ultra-low column bleed, extended column lifetimes, and excellent peak shape. The column utilizes MaxPeak High Performance Surface hardware technology, which reduces unwanted analyte/surface interactions that can lead to poor peak shape and signal intensity losses.
  • Provides complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Contains the most chemically stable phenyl sorbent on the market.
  • Offers ultra-low column bleed.
  • Ensures long column lifetimes.
  • Delivers excellent peak shape.
  • Utilizes MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions, preventing poor peak shape and signal intensity losses.
Catalog No. 50-288-6190 Supplier Waters Corp Supplier No. 186010664
This Premier BEH Phenyl VanGuard FIT Column provides complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. Its sorbent offers high chemical stability, ensuring ultra-low column bleed, extended column lifetimes, and excellent peak shape. The column utilizes MaxPeak High Performance Surface hardware technology, which reduces unwanted analyte/surface interactions that can lead to poor peak shape and signal intensity losses.
  • Provides complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Contains the most chemically stable phenyl sorbent on the market.
  • Offers ultra-low column bleed.
  • Ensures long column lifetimes.
  • Delivers excellent peak shape.
  • Utilizes MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions, preventing poor peak shape and signal intensity losses.