Learn More
Waters Corp Waters XBridge Premier BEH Phenyl Column, 2.5 μm, 4.6 mm X 150 mm
Supplier:  Waters Corp 186010351
Encompass_Preferred
XBridge Premier BEH Phenyl Columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. The sorbent is designed for chemical stability, ultra-low column bleed, and extended column lifetimes. These columns incorporate MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Complementary selectivity to C18 stationary phases
  • Chemically stable phenyl sorbent
  • Ultra-low column bleed
  • Long column lifetimes
  • Excellent peak shape
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions
Catalog No. 50-226-9359