Learn More
Waters Corp Waters Corporation Column, BEH Phenyl, 2.5 μm, 4.6 x 150 mm, VanGuard FIT
Catalog No. 502269362 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Premier columns provide advanced control over chromatographic separations. These columns are designed to mitigate the loss of metal-sensitive analytes through the innovative MaxPeak™ High Performance Surfaces (HPS) technology, which minimizes analyte/surface interactions, thereby improving analyte recovery, sensitivity, and reproducibility.
  • Reduced column conditioning and passivation times
  • Improved sensitivity and peak shapes
  • Simpler mobile phases
  • Time savings in method development
  • Reduced risk and greater confidence in data
  • Versatile application for small molecule, peptide, oligonucleotide, and glycan separations in both reversed-phase and HILIC modes
This item is not returnable. View return policy
Catalog No. 50-226-9362 Supplier Waters Corp Supplier No. 186010354
XBridge Premier columns provide advanced control over chromatographic separations. These columns are designed to mitigate the loss of metal-sensitive analytes through the innovative MaxPeak™ High Performance Surfaces (HPS) technology, which minimizes analyte/surface interactions, thereby improving analyte recovery, sensitivity, and reproducibility.
  • Reduced column conditioning and passivation times
  • Improved sensitivity and peak shapes
  • Simpler mobile phases
  • Time savings in method development
  • Reduced risk and greater confidence in data
  • Versatile application for small molecule, peptide, oligonucleotide, and glycan separations in both reversed-phase and HILIC modes