Learn More
Waters Corp GTxResolve Columns, BEH SEC, 450 Å, 2.5 μm, 7.8 × 300 mm
Catalog No. 502390726 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GTxResolve Premier BEH SEC 450 Å, 2.5 μm, 7.8 × 300 mm Column is designed to provide high resolution and fast size variant separations for AAVs and nucleic acids, helping to resolve uncertainty in gene therapy development.
  • Effectively resolves AAV monomers and aggregates due to its 450 Å pore size.
  • Features hydrophilic MaxPeak High Performance Surface hardware, which alleviates unwanted secondary interactions.
  • Promotes full sample recovery.
  • Supports robust, serotype-independent, and stable platform methods.
  • Efficiently packed 2.5 μm particles deliver industry-leading resolution without undesirable analyte shearing.
Catalog No. 50-239-0726 Supplier Waters Corp Supplier No. 186010587
The GTxResolve Premier BEH SEC 450 Å, 2.5 μm, 7.8 × 300 mm Column is designed to provide high resolution and fast size variant separations for AAVs and nucleic acids, helping to resolve uncertainty in gene therapy development.
  • Effectively resolves AAV monomers and aggregates due to its 450 Å pore size.
  • Features hydrophilic MaxPeak High Performance Surface hardware, which alleviates unwanted secondary interactions.
  • Promotes full sample recovery.
  • Supports robust, serotype-independent, and stable platform methods.
  • Efficiently packed 2.5 μm particles deliver industry-leading resolution without undesirable analyte shearing.