Learn More
Waters Corp SEC Column, 450 Å, 2.5 μm, 4.6 x 150 mm
Catalog No. 502390490 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This SEC column delivers high resolution and fast size variant separations for AAVs and nucleic acids. It resolves AAV monomers and aggregates using a 450 Å pore size, with hydrophilic MaxPeak High Performance Surface hardware preventing unwanted secondary interactions. Its efficiently packed 2.5 μm particles provide industry-leading resolution without analyte shearing.
  • High resolution and fast size variant separations for AAVs and nucleic acids.
  • Resolves AAV monomers and aggregates with 450 Å pore size.
  • MaxPeak High Performance Surface hardware prevents secondary interactions.
  • Efficiently packed 2.5 μm particles ensure industry-leading resolution.
  • Rugged BEH Technology particles offer inertness to ionic secondary interactions.
  • Manufactured under cGMP, ISO 9001 standards with stringent QC.
Catalog No. 50-239-0490 Supplier Waters Corp Supplier No. 186010584
This SEC column delivers high resolution and fast size variant separations for AAVs and nucleic acids. It resolves AAV monomers and aggregates using a 450 Å pore size, with hydrophilic MaxPeak High Performance Surface hardware preventing unwanted secondary interactions. Its efficiently packed 2.5 μm particles provide industry-leading resolution without analyte shearing.
  • High resolution and fast size variant separations for AAVs and nucleic acids.
  • Resolves AAV monomers and aggregates with 450 Å pore size.
  • MaxPeak High Performance Surface hardware prevents secondary interactions.
  • Efficiently packed 2.5 μm particles ensure industry-leading resolution.
  • Rugged BEH Technology particles offer inertness to ionic secondary interactions.
  • Manufactured under cGMP, ISO 9001 standards with stringent QC.