Learn More
Waters Corp XBridge Premier Oligonucleotide BEH C18 Column, 300 Å, 2.5 μm, 4.6 mm × 50 mm
Catalog No. 502392020 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier Oligonucleotide BEH C18 Column, 300 Å, 2.5 μm, 4.6 mm × 50 mm, is designed for the separation of oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography on UHPLC/HPLC systems. It incorporates BEH hybrid particle technology, providing unmatched pH and temperature stability. The column also features MaxPeak Premier high-performance surface technology, which ensures ultra-low non-specific adsorption.
  • Stationary phase batches are QC tested with the MassPREP Oligonucleotide Standard to guarantee consistent lot-to-lot performance.
  • Offers unmatched sensitivity, excellent reproducibility, and superior out-of-the-box performance.
  • 300Å pore size columns provide enhanced resolution for longer oligonucleotides.
Catalog No. 50-239-2020 Supplier Waters Corp Supplier No. 186010545
Add to Cart
Add to Cart
The XBridge Premier Oligonucleotide BEH C18 Column, 300 Å, 2.5 μm, 4.6 mm × 50 mm, is designed for the separation of oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography on UHPLC/HPLC systems. It incorporates BEH hybrid particle technology, providing unmatched pH and temperature stability. The column also features MaxPeak Premier high-performance surface technology, which ensures ultra-low non-specific adsorption.
  • Stationary phase batches are QC tested with the MassPREP Oligonucleotide Standard to guarantee consistent lot-to-lot performance.
  • Offers unmatched sensitivity, excellent reproducibility, and superior out-of-the-box performance.
  • 300Å pore size columns provide enhanced resolution for longer oligonucleotides.