Learn More
Waters Corp XBridge Premier Oligonucleotide BEH C18 Column, 300 Å, 2.5 μm, 2.1 × 150 mm
Catalog No. 502391736 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier Oligonucleotide BEH C18 Column, 300 Å, 2.5 μm, 2.1 × 150 mm, separates oligonucleotides via ion-pairing reversed-phase (IP-RP) chromatography on UHPLC/HPLC systems. It features BEH hybrid particle technology for pH and temperature stability and MaxPeak Premier high performance surface technology for ultra-low non-specific adsorption.
  • QC tested using MassPREP Oligonucleotide Standard to ensure lot-to-lot performance.
  • Provides unmatched sensitivity, exquisite reproducibility, and excellent out-of-the-box performance.
  • Enhanced resolution for longer oligonucleotides with 300Å pore size.
  • Suitable for short-mer oligonucleotide separations with 130Å pore size particle options.
  • Applicable in oligonucleotide and gene therapeutics.
Catalog No. 50-239-1736 Supplier Waters Corp Supplier No. 186010544
The XBridge Premier Oligonucleotide BEH C18 Column, 300 Å, 2.5 μm, 2.1 × 150 mm, separates oligonucleotides via ion-pairing reversed-phase (IP-RP) chromatography on UHPLC/HPLC systems. It features BEH hybrid particle technology for pH and temperature stability and MaxPeak Premier high performance surface technology for ultra-low non-specific adsorption.
  • QC tested using MassPREP Oligonucleotide Standard to ensure lot-to-lot performance.
  • Provides unmatched sensitivity, exquisite reproducibility, and excellent out-of-the-box performance.
  • Enhanced resolution for longer oligonucleotides with 300Å pore size.
  • Suitable for short-mer oligonucleotide separations with 130Å pore size particle options.
  • Applicable in oligonucleotide and gene therapeutics.