Learn More
Waters Corp Column, 300 Å, 2.5 μm, 2.1 x 100 mm, C18
Catalog No. 502392771 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier Oligonucleotide BEH C18 Column separates oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography on UHPLC/HPLC systems. It utilizes BEH hybrid particle technology for exceptional pH/temperature stability and MaxPeak Premier high-performance surface technology for ultra-low non-specific adsorption.
  • Separates oligonucleotides via ion-pairing reversed-phase (IP-RP) chromatography.
  • Incorporates BEH hybrid particle technology for pH and temperature stability.
  • Features MaxPeak Premier high performance surface technology for ultra-low non-specific adsorption.
  • Provides superior sensitivity and reproducibility.
  • QC tested for consistent lot-to-lot performance.
  • New 300 Å pore size columns enhance resolution for longer oligonucleotides.
  • Digital Certificates of Analysis (COAs) available online.
Catalog No. 50-239-2771 Supplier Waters Corp Supplier No. 186010543
The XBridge Premier Oligonucleotide BEH C18 Column separates oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography on UHPLC/HPLC systems. It utilizes BEH hybrid particle technology for exceptional pH/temperature stability and MaxPeak Premier high-performance surface technology for ultra-low non-specific adsorption.
  • Separates oligonucleotides via ion-pairing reversed-phase (IP-RP) chromatography.
  • Incorporates BEH hybrid particle technology for pH and temperature stability.
  • Features MaxPeak Premier high performance surface technology for ultra-low non-specific adsorption.
  • Provides superior sensitivity and reproducibility.
  • QC tested for consistent lot-to-lot performance.
  • New 300 Å pore size columns enhance resolution for longer oligonucleotides.
  • Digital Certificates of Analysis (COAs) available online.