Learn More
Waters Corp Oligonucleotide Column, C18, 130 Å, 2.5 μm
Catalog No. 502390916 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Premier Oligonucleotide BEH C18 Columns are engineered for the separation of synthetic oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography. These columns feature an ultra-low non-adsorptive surface from the MaxPeak Premier line, which significantly enhances sensitivity and reproducibility for metal-sensitive analytes like oligonucleotides.
  • Undergoes quality control testing to ensure consistent performance.
  • Includes a fully integrated and easily replaceable 5 mm pre-column.
  • Pre-column protects and extends the life of the analytical column without adding dispersion or impacting resolution.
  • VanGuard FIT Cartridge replacements contain the same qualified sorbent and are easily replaceable.
Catalog No. 50-239-0916 Supplier Waters Corp Supplier No. 186010689
XBridge Premier Oligonucleotide BEH C18 Columns are engineered for the separation of synthetic oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography. These columns feature an ultra-low non-adsorptive surface from the MaxPeak Premier line, which significantly enhances sensitivity and reproducibility for metal-sensitive analytes like oligonucleotides.
  • Undergoes quality control testing to ensure consistent performance.
  • Includes a fully integrated and easily replaceable 5 mm pre-column.
  • Pre-column protects and extends the life of the analytical column without adding dispersion or impacting resolution.
  • VanGuard FIT Cartridge replacements contain the same qualified sorbent and are easily replaceable.