Learn More
Waters Corp Waters Oligonucleotide Column, BEH C18, 2.5 μm, 2.1 x 150 mm
Supplier:  Waters Corp 186009838
Encompass_Preferred
This column separates detritylated synthetic oligonucleotides using ion-pair, reversed-phase chromatography. Rigorous QC testing with MassPrep OST standards ensures consistent performance for challenging oligonucleotide assays. As a MaxPeak Premier Class Column, it enhances reproducibility, peak shape, and recovery for metal-sensitive analytes. Broad pH compatibility (1 to 12) allows versatile use.
  • Separates detritylated synthetic oligonucleotides using ion-pair, reversed-phase chromatography.
  • QC tested with MassPrep OST standards for repeatable performance in demanding oligonucleotide assays.
  • Offers enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes as part of the MaxPeak Premier Class.
  • Provides broad pH compatibility and operational efficiency across a pH range of 1 to 12.
Catalog No. 50-222-4257