Learn More
Waters Corp Waters Corporation Column, XBridge Premier, C18, 2.5 μm, 2.1 mm ID
Supplier:  Waters Corp 186009837
Encompass_Preferred
The XBridge Premier Oligonucleotide BEH C18 Column separates detritylated synthetic oligonucleotides via ion-pair, reversed-phase chromatography. A MaxPeak Premier Class Column, it enhances reproducibility, peak shape, and recovery for metal-sensitive analytes, reducing column passivation and conditioning, and improving data quality.
  • QC tested with MassPrep OST standards for repeatable performance.
  • Reduces column passivation and conditioning.
  • Utilizes Ethylene-Bridged Hybrid (BEH) particle technology for stable oligonucleotide separations.
  • Offers mechanical strength and chemical stability.
  • Incorporates MaxPeak High-Performance Surface Technology to reduce analyte-surface interactions, enhancing recovery, sensitivity, and reproducibility.
  • Simplifies mobile phases by removing complex additives.
Catalog No. 50-222-4256